Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethylene glycol dimethyl ether, ≥99.7% (GC), MilliporeSigma™ Supelco™

Catalog No. 111019160
Click to view available options
Quantity:
1 mL
5 mL

Diethylene glycol dimethyl ether is a combustible liquid, which can be used in the synthesis of films, chemically similar to polyethylene glycol on low-density polyethylene and Si(100) surfaces via plasma polymerization and deposition.

Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

Ignition Point 370°F
CAS 111-96-6
Vapor Pressure 3 mmHg (20°C)
Vapor Density 4.6 (vs. air)
Density 0.944 g/mL at 20°C; 0.939 g/mL (at 25°C)
Flash Point 57°C
Linear Formula (CH3 OCH2 CH2)2 O
MDL Number MFCD00008503
Melting Point -64°C
Boiling Point 162°C
Synonym 2-Methoxyethyl ether; Bis(2-methoxyethyl) ether; Dimethyldiglycol; ′Diglyme’
Type Analytical Standard
Quantity 5 mL
Formula Weight 134.17
Percent Purity ≥99.7% (GC)
Grade Analytical Standard
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.