Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethylene Glycol Isopropyl Methyl Ether 98.0+%, TCI America™
SDP

Catalog No. D467725ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D467725ML 25 mL
D4677500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D467725ML Supplier TCI America Supplier No. D467725ML

Chemical Identifiers

CAS 77078-22-9
Molecular Formula C8H18O3
Molecular Weight (g/mol) 162.229
MDL Number MFCD28009631
InChI Key RJBIZCOYFBKBIM-UHFFFAOYSA-N
Synonym 2-[2-(2-Methoxyethoxy)ethoxy]propane
PubChem CID 3018692
IUPAC Name 2-[2-(2-methoxyethoxy)ethoxy]propane
SMILES CC(C)OCCOCCOC

Specifications

Color Colorless
Quantity 25 mL
Formula Weight 162.23
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethylene Glycol Isopropyl Methyl Ether
EINECSNumber (7)-1321
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.