Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethylene Glycol Mono(2-propyn-1-yl) Ether 97.0+%, TCI America™
SDP

Catalog No. D45805G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D45805G 5 g
D45801G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D45805G Supplier TCI America Supplier No. D45805G

Chemical Identifiers

CAS 7218-43-1
Molecular Formula C7H12O3
Molecular Weight (g/mol) 144.17
MDL Number MFCD00053353
InChI Key HUSDTFBXUYBZJD-UHFFFAOYSA-N
Synonym 2-[2-(2-Propyn-1-yloxy)ethoxy]ethanol
PubChem CID 81639
IUPAC Name 2-(2-prop-2-ynoxyethoxy)ethanol
SMILES C#CCOCCOCCO

Specifications

Color Yellow
Boiling Point 81°C
Quantity 5 g
Formula Weight 144.17
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethylene Glycol Mono(2-propyn-1-yl) Ether
EINECSNumber (2)-2398
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.