Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethylene glycol monomethyl ether, 98%

Catalog No. p-7023032
Click to view available options
Quantity:
500 mL
2500 mL

CAS: 111-77-3 | C5H12O3 | 120.15 g/mol

Diethylene glycol monomethyl ether is an industrial solvent and is also commonly used as a Fuel System Icing Inhibitor (FSII) in jet fuels.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethylene glycol monomethyl ether is an industrial solvent and is also commonly used as a Fuel System Icing Inhibitor (FSII) in jet fuels.

Solubility
Fully miscible in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is hygroscopic in nature. Store under dry inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-77-3
Molecular Formula C5H12O3
Molecular Weight (g/mol) 120.15
MDL Number MFCD00002871,MFCD00084416
InChI Key SBASXUCJHJRPEV-UHFFFAOYSA-N
Synonym 2-2-methoxyethoxy ethanol, diethylene glycol monomethyl ether, methyl carbitol, methyl digol, methoxydiglycol, ethanol, 2-2-methoxyethoxy, methyl dioxitol, diethylene glycol methyl ether, dowanol dm, poly-solv dm
PubChem CID 8134
ChEBI CHEBI:44836
IUPAC Name 2-(2-methoxyethoxy)ethanol
SMILES COCCOCCO

Specifications

Melting Point -70°C
Density 1.01
Boiling Point 194°C
Quantity 500 mL
Flash Point 83°C (181°F)
Odor Light
Refractive Index 1.426
Linear Formula CH3OCH2CH2OCH2CH2OH
Beilstein 1697812
Merck Index 14,6037
Solubility Information Fully miscible in water.
Formula Weight 120.15
Percent Purity 98%
Chemical Name or Material Diethylene glycol monomethyl ether, stabilized with 50 to 150mg/kg BHT
Show More Show Less
Hazard Category H227-H311-H319-H361d
Hazard Statement GHS H Statement
H361-H227
Suspected of damaging fertility or the unborn child.
Combustible liquid.
Precautionary Statement P201-P202-P210-P233-P240-P243-P264b-P280-P302+P352-P305+P351+P338-P308+P313-P312-P362-P374-P501c
EINECSNumber 203-906-6
RTECSNumber KL6125000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.