Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethylene Glycol Monovinyl Ether (stabilized with KOH) 96.0+%, TCI America™
SDP

Catalog No. D262325ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D262325ML 25 mL
D2623100ML 100 mL
D2623500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D262325ML Supplier TCI America Supplier No. D262325ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 929-37-3
Molecular Formula C6H12O3
Molecular Weight (g/mol) 132.16
MDL Number MFCD00192027
InChI Key WULAHPYSGCVQHM-UHFFFAOYSA-N
Synonym Vinyl Carbitol, 2-(2-Vinyloxyethoxy)ethanol
PubChem CID 13581
IUPAC Name 2-[2-(ethenyloxy)ethoxy]ethan-1-ol
SMILES OCCOCCOC=C

Specifications

Color Colorless
Boiling Point 96°C
Quantity 25 mL
Formula Weight 132.16
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethylene Glycol Monovinyl Ether (stabilized with KOH)
EINECSNumber (7)-0097
RTECSNumber KM5495500
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.