Learn More
Diethylenetriamine, 99%
Description
Diethylenetriamine is used as a solvent for plastics, dyes and in chemical synthesis. It serves as a curing agent for epoxy-resins. As a tridentate ligand, it form complexes such as Co(dien)(NO2)3 in coordination chemistry. It is also used as a fuel additive. It reacts with fatty acid resulting in the formation of amidoamines, which are used as corrosion inhibitors in the petroleum industry. It is associated with unsymmetrical dimethylhydrazine to prepare hydyne, which is a propellant for liquid-fuel rockets.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Diethylenetriamine is used as a solvent for plastics, dyes and in chemical synthesis. It serves as a curing agent for epoxy-resins. As a tridentate ligand, it form complexes such as Co(dien)(NO2)3 in coordination chemistry. It is also used as a fuel additive. It reacts with fatty acid resulting in the formation of amidoamines, which are used as corrosion inhibitors in the petroleum industry. It is associated with unsymmetrical dimethylhydrazine to prepare hydyne, which is a propellant for liquid-fuel rockets.
Solubility
Miscible with water and ethanol. Immiscible with ether.
Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, strong acids and copper.

Specifications
Specifications
Melting Point | -40°C |
Density | 0.952 |
Boiling Point | 200°C to 204°C |
Flash Point | 94°C (201°F) |
Odor | Ammonia-like |
Refractive Index | 1.483 |
Linear Formula | H2NCH2CH2NHCH2CH2NH2 |
Quantity | 2500 mL |
UN Number | UN2079 |
Beilstein | 605314 |
Show More |
Safety and Handling
Hazard Category | H227-H302+H312-H314-H317-H330-H335 |
Hazard Statement | GHS H Statement H314-H302-H312-H317 Causes severe skin burns and eye damage. Harmful if swallowed. Harmful in contact with skin. May cause an allergic skin reaction. |
Precautionary Statement | P210-P235-P260-P264b-P270-P271-P272-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: DIETHYLENETRIAMINE |
EINECSNumber | 203-865-4 |
RTECSNumber | IE1225000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.