Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethylenetriaminepentaacetic Acid Calcium Trisodium Salt Hydrate 98.0+%, TCI America™
SDP

Catalog No. D186925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D186925G 25 g
D18695G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D186925G Supplier TCI America Supplier No. D186925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 207226-35-5
Molecular Formula C14H20CaN3Na3O11
Molecular Weight (g/mol) 515.37
MDL Number MFCD00150947
InChI Key AWBWYUNJDQLQGT-UHFFFAOYSA-I
Synonym Calcium Trisodium Diethylenetriaminepentaacetate
PubChem CID 17749130
IUPAC Name calcium trisodium 2-[bis({2-[bis(carboxylatomethyl)amino]ethyl})amino]acetate hydrate
SMILES O.[Na+].[Na+].[Na+].[Ca++].[O-]C(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CCN(CC([O-])=O)CC([O-])=O

Specifications

Color White
Quantity 25 g
Formula Weight 497.36
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Diethylenetriaminepentaacetic Acid Calcium Trisodium Salt Hydrate
RTECSNumber MB8210000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.