Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethylgermanium dichloride

Catalog No. AA3410806
Click to view available options
Quantity:
5 g
25 g

As an intermediate | CAS: 13314-52-8 | C4H14Cl2Ge | 205.69 g/mol

As an intermediate

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13314-52-8
Molecular Formula C4H14Cl2Ge
Molecular Weight (g/mol) 205.69
MDL Number MFCD00013589
InChI Key FVUWAWRMMUJVGO-UHFFFAOYSA-N
Synonym diethylgermanium dichloride, diethyldichlorogermane, diethyldichlorogermanium, dichloro diethyl germane, germane,dichlorodiethyl, acmc-1c1bs, diethylgermanium chloride, dichlorodiethylgermane
PubChem CID 83334
IUPAC Name dichloro(diethyl)germane
SMILES Cl.Cl.CC[GeH2]CC

Specifications

Melting Point -37°C
Color Colorless
Density 1.372 g/mL
Boiling Point 175°C
Flash Point 63°C (145°F)
Odor Acrid
Refractive Index 1.471
Quantity 5 g
UN Number UN3265
Sensitivity Moisture sensitive
Solubility Information Decomposes in water and NH3; Soluble in organic solvents
Formula Weight 201.64
Physical Form Liquid
Chemical Name or Material Diethylgermanium dichloride
Show More Show Less
Hazard Category H227-H302+H312+H332-H314-H335
Hazard Statement GHS H Statement
H314-H318-H302-H312-H332-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Combustible liquid.
Precautionary Statement P210-P235-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 236-344-5
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.