Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethylisopropylsilyl Trifluoromethanesulfonate 98.0+%, TCI America™
SDP

Catalog No. T14681G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T14681G 1 g
T14685G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T14681G Supplier TCI America Supplier No. T14681G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 126889-55-2
Molecular Formula C8H17F3O3SSi
Molecular Weight (g/mol) 278.361
MDL Number MFCD00191689
InChI Key RIXRWPWLOMOUPT-UHFFFAOYSA-N
Synonym Diethylisopropylsilyl Triflate, Trifluoromethanesulfonic Acid Diethylisopropylsilyl Ester
PubChem CID 10956833
IUPAC Name [diethyl(propan-2-yl)silyl] trifluoromethanesulfonate
SMILES CC[Si](CC)(C(C)C)OS(=O)(=O)C(F)(F)F

Specifications

Color Yellow
Quantity 1 g
UN Number 1760
Formula Weight 278.36
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material Diethylisopropylsilyl Trifluoromethanesulfonate
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.