Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethylstilbestrol, 99%

Catalog No. p-110011
Click to view available options
Quantity:
1 g
5 g
10 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 56-53-1 | C18H20O2 | 268.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-53-1
Molecular Formula C18H20O2
Molecular Weight (g/mol) 268.36
MDL Number MFCD00002373
InChI Key RGLYKWWBQGJZGM-ISLYRVAYSA-N
Synonym diethylstilbestrol, stilbestrol, stilboestrol, distilbene, agostilben, estrobene, estromenin, stilboestroform, antigestil, palestrol
PubChem CID 448537
ChEBI CHEBI:41922
SMILES CC\C(=C(\CC)C1=CC=C(O)C=C1)C1=CC=C(O)C=C1

Specifications

Melting Point 169°C to 175°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Beilstein 06, III, 5619
Merck Index 15, 3148
Solubility Information Solubility in water: practically insoluble in water. Other solubilities: soluble in ethylether,ethanol,chloroform,aceton,e,methanol,arachis oil,olive oil
IUPAC Name 4-[(3E)-4-(4-hydroxyphenyl)hex-3-en-3-yl]phenol
Formula Weight 268.34
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Diethylstilbestrol, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Suspected of causing cancer.
May damage the unborn child.
Precautionary Statement GHS P Statement:
Use personal protective equipment as required.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 200-278-5
RTECSNumber WJ5600000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.