Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Diethylstilbestrol Monoclonal Antibody (2E6)
GREENER_CHOICE

Catalog No. PIMA549709
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA549709 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA549709 Supplier Invitrogen™ Supplier No. MA549709
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Antibody tested in indirect ELISA.

Diethylstilbestrol is a synthetic, nonsteroidal form of estrogen. A well-known teratogen and carcinogen, diethylstilbestrol inhibits the hypothalamic-pituitary-gonadal axis, thereby blocking the testicular synthesis of testosterone, lowering plasma testosterone, and inducing a chemical castration. Diethylstilbestrol is used in the treatment of menopausal and postmenopausal disorders. It was also used formerly as a growth promoter in animals. [National Center for Biotechnology Information. PubChem Compound Database; CID=448537].
TRUSTED_SUSTAINABILITY

Specifications

Antigen Diethylstilbestrol
Applications ELISA, Immunohistochemistry, Lateral Flow, SDS-Page
Classification Monoclonal
Clone 2E6
Concentration 2.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene Alias C18H20O2; Diethylstilbestrol; Distilbene; Stilbestrol; Stilboestrol
Host Species Mouse
Immunogen Conjugate of DES hapten and KLH.
Purification Method Protein G
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.