Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethylzinc, 1.5M solution in toluene, AcroSeal™, ACROS Organics™

Click to view available options
Quantity:
100mL
800mL
Packaging:
AcroSeal™ Glass Bottle
557-20-0,108-88-3
C4H10Zn
123.504
MFCD00009021
IPSRAFUHLHIWAR-UHFFFAOYSA-N
diethylzinc,zinc, diethyl,diethyl zinc,zinc ethide,zinc ethyl,unii-s0w5nqh7c6,et2zn,znet2,s0w5nqh7c6,c2h5 2zn
11185
zinc;ethane
C[CH2-].C[CH2-].[Zn+2]
557-20-0,108-88-3
C4H10Zn
123.504
MFCD00009021
IPSRAFUHLHIWAR-UHFFFAOYSA-N
diethylzinc,zinc, diethyl,diethyl zinc,zinc ethide,zinc ethyl,unii-s0w5nqh7c6,et2zn,znet2,s0w5nqh7c6,c2h5 2zn
11185
zinc;ethane
C[CH2-].C[CH2-].[Zn+2]
Linear Formula (C2H5)2Zn
Concentration 1.4 to 1.6M
Density 0.9130g/mL
Merck Index 15, 3151
Formula Weight 123.5
Physical Form Liquid
Packaging AcroSeal™ Glass Bottle
Solubility Solubility in water: reacts violently. Other solubilities: miscible with most hydrocarbons
Specific Gravity 0.913
Color Colorless
Quantity 800mL
Chemical Name or Material Diethylzinc, 1.5M Solution in Toluene
Show More Show Less

GHS H Statement
Highly flammable liquid and vapor. In contact with water releases flammable gases which may ignite spontaneously. May be fatal if swallowed and enters airways. Causes severe skin burns and eye damage. May cause drowsiness or dizziness. Suspected of damaging the unborn child. May cause damage to organs through prolonged or repeated exposure. Toxic to aquatic life with long lasting effects. Reacts violently with water.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: Rinse mouth. Do NOT induce vomiting. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Store in a dry place. Store in a closed container.

Danger