Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethylzinc, 96%

Catalog No. aa3956422
Click to view available options
Quantity:
100 g
Packaging:
Cylinder

CAS: 557-20-0 | C4H10Zn | 123.50 g/mol

It is used as an important reagent in organic chemistry. It is used in organic synthesis as a source of the ethyl carbanion in addition reactions to carbonyl groups.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 557-20-0
Molecular Formula C4H10Zn
Molecular Weight (g/mol) 123.50
MDL Number MFCD00009021
InChI Key HQWPLXHWEZZGKY-UHFFFAOYSA-N
Synonym diethylzinc, zinc, diethyl, diethyl zinc, zinc ethide, zinc ethyl, unii-s0w5nqh7c6, et2zn, znet2, s0w5nqh7c6, c2h5 2zn
PubChem CID 11185
IUPAC Name zinc;ethane
SMILES CC[Zn]CC

Specifications

Melting Point -28°C
Color Colorless
Density 1.18 g/mL
Boiling Point 124°C
Flash Point -18°C (-1°F)
Packaging Cylinder
Linear Formula Zn(C2H5)2
Quantity 100 g
UN Number UN3394
Sensitivity Air and moisture sensitive
Merck Index 14,3131
Solubility Information Reacts with water.
Formula Weight 123.49
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material Diethylzinc
Show More Show Less
Hazard Category H225-H250-H260-H314-H335
Hazard Statement GHS H Statement
H250-H260-H314-H318
Catches fire spontaneously if exposed to air.
In contact with water releases flammable gases which may ignite spontaneously.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P210-P222-P223-P231+P232-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P302+P334-P304+P340-P305+P351+P338-P310-P335+P334-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.2; Packing Group: I; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, PYROPHORIC, WATER-REACTIVE
EINECSNumber 209-161-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.