Alfa Aesar™ Diethylzinc, nominally 15% w/w in hexanes, packaged under Argon in resealable ChemSeal™ bottles

Manufacturer: Alfa Aesar™ 4488022

Quantity 100g

 View more versions of this product

Catalog No. AA4488022

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 557-20-0
    Molecular Formula C4H10Zn
    Linear Formula Zn(C2H5)2
    Formula Weight 123.49
    MDL Number MFCD00009021
    UN Number UN3394
    Chemical Name or Material Diethylzinc

    Specifications

    Formula Weight 123.49
    Health Hazard 2 GHS H Statement
    H225-H250-H260-H252-H304-H361-H373-H314-H336
    Highly flammable liquid and vapor.
    Catches fire spontaneously if exposed to air.
    In contact with water releases flammable gases which may ignite spontaneously.
    Self-heating in large quantities
    may catch fire.
    May be fatal if swallowed and enters airways.
    Suspected of damaging fertility or the unborn child.
    May cause damage to organs through prolonged or repeated exposure.
    Causes severe skin burns and eye damage.
    May cause drowsiness or dizziness.
    Health Hazard 3 GHS P Statement
    P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P422a-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Store contents under inert gas.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 557-20-0
    Sensitivity Air & Moisture Sensitive
    Health Hazard 1 Danger
    Flash Point −40°C (−40°F)
    Storage Note 1
    • Packaged under Argon
    Color Colorless to pale brown
    Packaging ChemSeal™ bottles
    Physical Form Liquid
    Molecular Formula C4H10Zn
    Linear Formula Zn(C2H5)2
    MDL Number MFCD00009021
    UN Number UN3394
    Quantity 100g
    Density 0.720g/mL
    Concentration Nominally 15% w/w in hexanes
    TSCA TSCA
    DOT Information Transport Hazard Class: 4.2; Packing Group: I; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, PYROPHORIC, WATER-REACTIVE
    Chemical Name or Material Diethylzinc

    GHS H Statement
    H225-H250-H260-H252-H304-H361-H373-H314-H336
    Highly flammable liquid and vapor.
    Catches fire spontaneously if exposed to air.
    In contact with water releases flammable gases which may ignite spontaneously.
    Self-heating in large quantities
    may catch fire.
    May be fatal if swallowed and enters airways.
    Suspected of damaging fertility or the unborn child.
    May cause damage to organs through prolonged or repeated exposure.
    Causes severe skin burns and eye damage.
    May cause drowsiness or dizziness.

    GHS P Statement
    P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P422a-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Store contents under inert gas.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    • Packaged under Argon

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit