Learn More
Stoelting™ Dietrich Bulldog Clamp
SDP
Stoelting™ Dietrich Bulldog Clamp is useful in surgeries that require precise blood stoppage.
$394.00
Specifications
For Use With (Application) Neuroscience
Length (English) 2 in.
View More Specs
Products 2
Catalog Number Mfr. No. Model Price Quantity & Availability  
Catalog Number Mfr. No. Model Price Quantity & Availability  
10-000-479
SDP
View Documents
Stoelting™
5212038P
Straight
Each for $394.00
Only null left
Add to Cart
10-000-480
SDP
View Documents
Stoelting™
5212039P
Curved
Each for $394.00
Only null left
Add to Cart
Description
Description
One of the most common applications where a bulldog clamp can be used is during coronary surgery where blood flow must be stopped. In general the bulldog clamp, with its many angled varieties and surgical applications, is an essential tool during surgery where blood flow to an organ must be stopped or controlled.
Specifications
Specifications
Neuroscience
2 in.
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.