Learn More
BD Difco™ Dehydrated Culture Media: Fluid Thioglycollate Medium
Shop All BD Diagnostic Systems ProductsDescription
- Formulation: Pancreatic digest of Casein 15g/L, Yeast extract 5g/L, Dextrose 5.5g/L, Sodium Chloride 2.5g/L, L-Cystine 0.5g/L, Sodium Thioglycollate 0.5g/L, Agar 0.75g/L, Resazurin 1mg/L
Certifications
AOAC, BAM, BS12, COMPF, EP, USDA, USP
Specifications
Specifications
| Certifications/Compliance | AOAC, BAM, BS12, COMPF, EP, USDA, USP |
| Content And Storage | 2°C to 8°C |
| Form | Dehydrated |
| Media Type | Fluid Thioglycollate Medium |
| Product Type | Dehydrated Culture Media |
| Formulation | Pancreatic digest of Casein 15g/L, Yeast extract 5g/L, Dextrose 5.5g/L, Sodium Chloride 2.5g/L, L-Cystine 0.5g/L, Sodium Thioglycollate 0.5g/L, Agar 0.75g/L, Resazurin 1mg/L |
| For Use With (Application) | Used for sterility testing |
| Packaging Type | Pail |
| Quantity | 2000 g |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.