Learn More
Specifications
Specifications
| Certifications/Compliance | AOAC, BAM, CCAM, COMPF, ISO, SMWW, USDA |
| Content And Storage | <30°C |
| Form | Dehydrated |
| Media Type | Nutrient Agar |
| Product Type | Nutrient Agar |
| Formulation | Beef extract 3g/L, Peptone 5g/L, Agar 15g/L |
| For Use With (Application) | General purpose medium for the cultivation of a wide variety of microorganisms |
| Packaging Type | Bottle |
| Quantity | 100 g |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.