Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Dehydrated Culture Media: Potato Dextrose Agar

Catalog No. DF0013158 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
100 g
500 g
2000 g
Packaging Type:
Bottle
Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging Type
DF0013-15-8 100 g Bottle
DF0013-07-8 2000 g Pail
DF0013-17-6 500 g Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. DF0013-15-8 Supplier BD Supplier No. 213300
This item is not returnable. View return policy

For cultivation of Yeasts and molds. BD Difco™ Dehydrated Culture Media: Potato Dextrose Agar is supplied in dehydrated format.

  • Dehydrated format
  • Culture media
  • DNR (Latex) free
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS, 11014, AOAC, BAM, BS12, CCAM, CMPH2, COMPF, EP, JP, MCM9, SMD, USP
Content And Storage 2°C to 30°C
Form Dehydrated
Media Type Potato Dextrose Agar
Product Type Agar
Formulation Potato Starch (from infusion) 4g/L, Dextrose 20g/L, Agar 15g/L
For Use With (Application) Isolation of Yeasts and molds
Packaging Type Bottle
Quantity 100 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.