Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Glycerol

Catalog No. p-4888799
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Used as a fixative, in bacterial preservation media and in the isolation and cultivation of many organisms. BD™ Difco™ Glycerol is a highly purified alcohol that helps support the growth of a wide variety of microorganisms, including mycobacterial species and fastidious bacteria.

Glycerol is a viscous, highly purified, fermentable alcohol often used to supplement the base of culture media (e.g. Agar). Glycerol is mixed into the solution with the culture base to support the growth of a wide variety of microorganisms.

  • CAS Number: 56-81-5
  • EC number: 200-289-5
  • Chemical formula: C3H8O3
  • Molar mass: 92.09g/mol
  • Density: 1.261 g/cm3
  • Solubility in water: miscible
  • Viscosity: 1.412 Pa•s
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS, 11014
Color Colorless
Content And Storage 15°C to 30°C
Additive Glycerol
Form Highly Viscous
For Use With (Application) Bacterial preservation media and in the isolation and cultivation of many organisms
Packaging Type Bottle
Quantity 500 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.