Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva DIGE Gel and DIGE Buffer Kit

Catalog No. 45002177 Shop All Cytiva Products
Change view
Click to view available options
Color:
Blue
Colorless
Form:
Liquid
Solid
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Form
45-002-177 Blue Liquid
45-002-178 Colorless Solid
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 45-002-177 Supplier Cytiva Supplier No. 28937452
NLSU: Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Kit: 2 x 125mL anode buffer; 4 x 125mL cathode buffer; 12 tubes agarose sealing solution

Tools for increasing the quality, reproducibility, and convenience of 2-D DIGE using Ettan™ DIGE System

  • DIGE Gel run with DIGE Buffer Kit results in spot maps with reproducibility compared to homecast polyacrylamide gels, thereby reducing technical variation between gels
  • Ready-to-use
  • Comes with long shelf-life
  • Eliminates the need to make buffers, cast gels and wash glass plates
  • Offers quality and performance of the Laemmli system with reproducibility, convenience and time savings
DIGE Gel
  • Should be used with the DIGE Buffer kit
  • 12.5% precast low fluorscent polyacrylamide gel
  • Cast in low fluorescent glass cassette developed for second dimension 2-D electrophoresis
  • Solid
  • Dimension: 10 x 7.7 x 0.03 in. (255 x 196 x 1mm)
DIGE Buffer Kit
  • Designed two-buffer system for running up to twelve DIGE Gels simultaneously
  • Contains the amount of reagents necessary for a single run of up to 12 DIGE Gels
  • Liquid

TRUSTED_SUSTAINABILITY

Specifications

Boiling Point 290°C
Description Specially designed two-buffer system for running up to twelve DIGE gels simultaneously
Color Blue
Form Liquid
For Use With (Application) For differential protein analysis
Kit Contents Running Buffers and sealing solution for attaching immobiline™ DryStrip gels (ipg strips) to the top of the Polyacrylamide Gel, Anode Buffer, cathode Buffer, 12 tubes Agarose sealing solution
Melting Temperature 18°C
Molecular Weight (g/mol) 92.09472
Quantity 1 Kit: 2 x 125 mL anode buffer; 4 x 125 mL cathode buffer; 12 tubes agarose sealing solution
Storage Requirements 2 - 8°C
Product Type DIGE Gel and Buffer Kit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.