Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Digestion Reagent, with Mercury Catalyst, Ricca Chemical
SDP

Catalog No. p-3251611
Click to view available options
Quantity:
1 L
4 L
Packaging:
Natural Poly Bottle
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.

For Analysis of Nitrogen (Kjeldahl)

Specifications

Chemical Name or Material Digestion Reagent
Quantity 1 L
Color Colorless
Name Note With Mercury Catalyst
CAS 7732-18-5, 7664-93-9, 7778-80-5, 21908-53-2
CAS Min % 65.0, 24.42, 8.45, 0.13
Packaging Natural Poly Bottle
CAS Max % 68.0, 25.42, 8.8, 0.14
Physical Form Liquid
Grade Laboratory
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.