Learn More
Techcyte Digital Gram Stain Analysis
Description
Digital Gram Analysis
- Solution is a revolutionary screening tool that helps medical technologists read bacteriology slides more efficiently and accurately
- AI-based algorithm, lab-optimized workflow, and LIS integration
- Used to identify various bacteria, cells, and yeasts in Gram stained slides.
Specifications
Specifications
| Product Type | Detection Software |
| For Use With (Application) | Analyzing Data |
| Operating System | Microsoft Windows 10 IoT 64-bit |
For investigational use only in the US.