Learn More
Quincy Lab Inc Digital Laboratory Gravity Convection Ovens
Easier to accurately set than analog ovens.
Supplier: Quincy Lab Inc 30GCE
Description
- Digital microprocessor control
- Gravity convection
- Durable double wall construction
- Scratch resistant hammer finish
- Corrosion resistant aluminized interior
- Made with UL and CSA recognized components
Specifications
| 2 cu. ft. | |
| 56.6 L | |
| 16 | |
| 18 Months | |
| 20 x 14 x 25 in., 51 x 36 x 64 cm | |
| 51 cm | |
| 36 cm | |
| 64 cm | |
| 115 V | |
| 10.5 A |
| CUL, IEC 61010-1, UL 61010-1 | |
| Digital Laboratory Oven | |
| ±0.5°C | |
| 30.5 x 45.7 x 39.6 cm | |
| 20 in. | |
| 14 in. | |
| 16 in. | |
| 70 lb. | |
| 1200 w |
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov