Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LaMotte™ Digital Tapwater Tour Kit



Click to view available options
Includes:
Intructor Manual, Student Manual, TesTab™ reagents and sample bags for 50 students
TesTab™ reagents and sample bags for 50 students
Description
- Instructor manual including background information, set up and shopping list
- Student Manual including test procedures, data sheets and puzzles
- Students work in groups and individually
- Works on computers or tablets
- Great for inclusion classrooms
- Refill includes reagents and sample bags for 50 students
Specifications
Specifications
Product Type | Curriculum and Hands-on Water Chemistry Tests |
Description | Digital Tapwater Tour Kit |
Grade | 4 to 8 |
Age | 9 to 14 |
Includes | Intructor Manual, Student Manual, TesTab™ reagents and sample bags for 50 students |
Material | Plastic sample bags, tableted TesTab™ reagents |
Quantity | 1 |
Class Size | 50 Students |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction