Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LaMotte™ Digital Tapwater Tour Kit
SDPSCI_ED

Catalog No. s72761
Click to view available options
Includes:
Intructor Manual, Student Manual, TesTab™ reagents and sample bags for 50 students
TesTab™ reagents and sample bags for 50 students

A classroom investigation of how and why the characteristics of tap water can vary and affect every day activities

  • Instructor manual including background information, set up and shopping list
  • Student Manual including test procedures, data sheets and puzzles
  • Students work in groups and individually
  • Works on computers or tablets
  • Great for inclusion classrooms
  • Refill includes reagents and sample bags for 50 students

Specifications

Product Type Curriculum and Hands-on Water Chemistry Tests
Description Digital Tapwater Tour Kit
Grade 4 to 8
Age 9 to 14
Includes Intructor Manual, Student Manual, TesTab™ reagents and sample bags for 50 students
Material Plastic sample bags, tableted TesTab™ reagents
Quantity 1
Class Size 50 Students
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.