Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell Digitcare™ Defender T™ DFR12-T Nitrile Exam Gloves

Catalog No. 19500951 Shop All Ansell Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-500-951 Medium
19-500-950 Small
19-500-952 Large
19-500-953 X-Large
19-500-954 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-500-951 Supplier Ansell Supplier No. DFR122T
This item is not returnable. View return policy

Specially designed for the rigors of tactical environments in a low-signature, military green color to effectively blend in with uniforms. Gloves have been tested against both fentanyl and gastric acid (vomit) to simulate hazardous, real-world overdose situations.

  • Specifically designed to meet the rigors of military and tactical environments
  • Gloves have been tested against fentanyl and gastric acid for advanced, real world overdose situations
  • Low-signature, tactical green color to match military uniforms
  • Delivers advanced barrier protection (AQL 0.65)
  • Non-stick surface resists duct tape and adhesives
  • Textured fingers for enhanced grip capabilities
  • Recommended for use as an outer-glove when double-gloving is necessary
TRUSTED_SUSTAINABILITY

Specifications

Chlorinated Yes
AQL 0.65
Color Green
Disposable Single-use
Exterior Finish Partially Textured
Finger Thickness (English) 8.3 mil
Finger Thickness (Metric) 0.21 mm
Latex Free Yes
Material Nitrile
Palm Thickness (English) 5.5 mil
Palm Thickness (Metric) 0.14 mm
Powdered Powder Free
Size Medium
Sterility Non-sterile
Thickness (English) 5.5 mil
Thickness (Metric) 0.14 mm
FDA 510 (k) No
Manufacturing/QMS Audit Standards ISO 13485, ISO 9001
Model DFR12-2T
Series Defender T™ DFR12-T
Length (English) 12 in.
Length (Metric) 30 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.