Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diglycolic anhydride, 97%

Catalog No. p-7027609
Click to view available options
Quantity:
10 g
50 g

CAS: 4480-83-5 | C4H4O4 | 116.072 g/mol

Diglycolic anhydride is used in the preparation of N,N-diethyldiglycollamsaeure by reacting with diethylamine. It is an useful bio chemical for proteomics research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diglycolic anhydride is used in the preparation of N,N-diethyldiglycollamsaeure by reacting with diethylamine. It is an useful bio chemical for proteomics research.

Solubility
Soluble in chloroform.

Notes
Moisture sensitive. Incompatible with strong acids, strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4480-83-5
Molecular Formula C4H4O4
Molecular Weight (g/mol) 116.072
MDL Number MFCD00006677
InChI Key PIYNUZCGMLCXKJ-UHFFFAOYSA-N
Synonym diglycolic anhydride, oxydiacetic anhydride, digylcolicanhydride, 4-maleimidobutyric acid hydrazide hydrochloride, r-1-methyl-2-pyrrolidineethanamine dihydrochloride, diglycolicanhydride, digylcolic anhydride, acetic acid, oxydi-, cyclic anhydride, p-dioxane-2,6-dione, pubchem24315
PubChem CID 78232
IUPAC Name 1,4-dioxane-2,6-dione
SMILES C1C(=O)OC(=O)CO1

Specifications

Melting Point 91°C to 94°C
Boiling Point 240°C to 241°C
Quantity 10 g
Beilstein 112527
Sensitivity Moisture sensitive
Solubility Information Soluble in chloroform.
Formula Weight 116.07
Percent Purity 0.97
Chemical Name or Material Diglycolic anhydride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 224-761-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.