Alfa Aesar™ Diglycolic anhydride, 97%

Manufacturer: Alfa Aesar™ B2449809

Quantity 10g

 View more versions of this product

Catalog No. AAB2449809

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 4480-83-5
    Molecular Formula C4H4O4
    Formula Weight 116.07
    MDL Number MFCD00006677
    Beilstein 112527
    Synonym 1,4-Dioxane-2,6-dione
    Chemical Name or Material Diglycolic anhydride

    Specifications

    Formula Weight 116.07
    MDL Number MFCD00006677
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Boiling Point 240° to 241°C
    Molecular Formula C4H4O4
    CAS 4480-83-5
    Beilstein 112527
    Synonym 1,4-Dioxane-2,6-dione
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Melting Point 91° to 94°C
    Quantity 10g
    Chemical Name or Material Diglycolic anhydride
    Assay 97%

    Description

    Applications
    Diglycolic anhydride is used in the preparation of N,N-diethyldiglycollamsaeure by reacting with diethylamine. It is an useful bio chemical for proteomics research.

    Solubility
    Soluble in chloroform.

    Notes
    Moisture sensitive. Incompatible with strong acids, strong oxidizing agents and strong bases.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit