Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diglycolic anhydride, 97%

Catalog No. p-111345
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle

CAS: 4480-83-5 | C4H4O4 | 116.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4480-83-5
Molecular Formula C4H4O4
Molecular Weight (g/mol) 116.07
MDL Number MFCD00006677
InChI Key PIYNUZCGMLCXKJ-UHFFFAOYSA-N
Synonym diglycolic anhydride, oxydiacetic anhydride, digylcolicanhydride, 4-maleimidobutyric acid hydrazide hydrochloride, r-1-methyl-2-pyrrolidineethanamine dihydrochloride, diglycolicanhydride, digylcolic anhydride, acetic acid, oxydi-, cyclic anhydride, p-dioxane-2,6-dione, pubchem24315
PubChem CID 78232
IUPAC Name 1,4-dioxane-2,6-dione
SMILES C1C(=O)OC(=O)CO1

Specifications

Melting Point 90°C to 95°C
Color Cream to White
Boiling Point 240°C to 241°C
Infrared Spectrum Authentic
Assay Percent Range 0.97
Packaging Glass bottle
Quantity 25 g
Beilstein 19, 153
Solubility Information Solubility in water: soluble
Formula Weight 116.07
Percent Purity 0.97
Physical Form Crystalline Powder, Crystals or Needles
Chemical Name or Material Diglycolic anhydride, 0.97
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER
EINECSNumber 224-761-5

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.