Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Digoxigenin, ≥98% (HPLC), MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco Products
Click to view available options
Quantity:
100 mg
500 mg
Description
Featured Industries: Environmental and Food and Beverages

Specifications
Specifications
CAS | 1672-46-4 |
Physical Form | Neat |
Linear Formula | C23H34O5 |
MDL Number | MFCD00871861 |
Melting Point | 222°C |
Synonym | 3 beta,12 beta,14 beta,21-Tetrahydroxy-20(22)-norcholenic acid lactone; 3 beta,12 beta,14-Trihydroxy-5 beta,20(22)-cardenolide; 5 beta,20(22)-Cardenolide-3 beta,12 beta,14-triol; Lanadigigenin |
Type | Analytical Standard |
Quantity | 100 mg |
Formula Weight | 390.51 |
Percent Purity | ≥98% (HPLC) |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction