Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Digoxigenin Antibody (21H8), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 3 publications
Supplier: Novus Biologicals NBP231191
Description
Digoxigenin Monoclonal specifically detects Digoxigenin in All Species samples. It is validated for Western Blot, ELISA, Immunohistochemistry, In-situ Hybridization, Southern Blot.Specifications
Digoxigenin | |
Monoclonal | |
LYOPH | |
Western Blot 1:500-1:5000, ELISA 1:1000 - 1:10000, Immunohistochemistry 1:200-1:2000, In-situ Hybridization, Southern Blot 1:500-1:5000 | |
Mouse | |
Protein A purified | |
RUO | |
Reacts with free and bound digoxigenin | |
All species | |
Purified |
Western Blot, ELISA, Immunohistochemistry, In Situ Hybridization (ISH), Southern Blot | |
21H8 | |
Unconjugated | |
DIG | |
Digoxigenin | |
100 μg | |
Primary | |
Reconstitute with 0.1 ml distilled water. | |
Store at 4C short term. Aliquot and store at -80C long term. Avoid freeze-thaw cycles. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction