Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Digoxigenin Antibody, Novus Biologicals™
SDP

Catalog No. p-7200859 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
0.5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017724 0.1 mL
NB10065495 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB017724 Supplier Novus Biologicals Supplier No. NB100654950.1ML
Only null left
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

Digoxigenin Polyclonal specifically detects Digoxigenin in Non-species specific samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Frozen, In-situ Hybridization.

Specifications

Antigen Digoxigenin
Applications In Situ Hybridization (ISH), Immunohistochemistry (Frozen), Immunohistochemistry, Western Blot, ELISA
Classification Polyclonal
Concentration 5.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, ELISA 1:100-1:5000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, In-situ Hybridization
Formulation Phosphate buffered saline with 0.09% Sodium Azide
Gene Alias DIG
Host Species Sheep
Immunogen Digoxin-3-CM conjugated to keyhole limpet haemocyanin
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes Digoxigenin, a steroid found in plants that is often attached to oligonucleotides and used as a molecular probe to detect mRNA production. This cross reacts with Digoxin and Digitoxin. Negligible reactivity has been observed with Ouabain.
Target Species Non-species specific
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.