Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Digoxigenin Antibody (DIG45) - VHH, Novus Biologicals™
SDP

Catalog No. NB408435 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB408435 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB408435 Supplier Novus Biologicals Supplier No. NBP306992
Only null left
Add to Cart
Add to Cart

Llama Monoclonal Antibody

Digoxigenin Monoclonal antibody specifically detects Digoxigenin in Plant samples. It is validated for Western Blot, ELISA, SDS-Page

Specifications

Antigen Digoxigenin
Applications Western Blot, ELISA, SDS-Page
Classification Monoclonal
Clone DIG45
Conjugate Unconjugated
Dilution Western Blot 1:1000, ELISA 1:2000, SDS-Page
Formulation 1X PBS, pH 7.4, 20% (v/v) Glycerol
Gene Alias DIG
Host Species Llama
Immunogen Digoxigenin
Purification Method Affinity purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Plant
Content And Storage Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.