Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Digoxigenin Monoclonal Antibody (21H8), QED Bioscience™
SDP

Catalog No. 502278287
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-227-8287 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-227-8287 Supplier Leinco Technologies Inc Supplier No. 90012100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Digoxigenin Monoclonal antibody specifically detects Digoxigenin in Chemical samples. It is validated for ELISA, Immunohistochemistry, Western Blot

Specifications

Antigen Digoxigenin
Applications ELISA, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 21H8
Concentration 0.5-1.0 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris with 0.1M glycine, 2% sucrose and no preservative
Gene Alias C23H34O5; DIG; Digitalis
Host Species Mouse
Immunogen Digoxigenin conjugate.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG1
Target Molecule Chemical
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.