Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Digoxigenin Monoclonal Antibody (Dig), Abbomax

Catalog No. 502204378
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-4378 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-4378 Supplier Abbomax Inc Supplier No. 605680
Add to Cart
Edge
Add to Cart

Mouse Monoclonal Antibody

Positive control:

Digoxigenin Monoclonal antibody specifically detects Digoxigenin in Chemical samples. It is validated for ELISA, Immunohistochemistry (Paraffin)

Specifications

Antigen Digoxigenin
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone Dig
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with proprietary stabilizer and 0.01% sodium azide
Gene Alias C23H34O5; DIG; Digitalis
Host Species Mouse
Immunogen A chmically conjugated Digoxigenin
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Target Molecule Chemical
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.