Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Digoxigenin Tyramide
GREENER_CHOICE

Catalog No. 7236100U
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
7236100U 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 7236100U Supplier Tocris Bioscience™ Supplier No. 7236/100U
Add to Cart
Add to Cart
This item is not returnable. View return policy

Reagent used for Tyramide Signal Amplification in IHC, ICC and ISH

Digoxigenin Tyramide is a hapten-labeled tyramide reagent used for Tyramide Signal Amplification (TSA) in immunohistochemistry (IHC), immunocytochemistry (ICC), and in situ hybridization (ISH). It uses horseradish peroxidase (HRP) to catalyze localized deposition of Digoxigenin Tyramide molecules. Digoxigenin Tyramide is suitable for either fluorescence or chromogenic detection.

Specifications

Quantity 100 μg
Target Tyramide Signaling Amplification/TSA
Purity 0.95
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.