Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Digoxin, 95%

Catalog No. ac228120010
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle

Catalog No. AC228120010

Supplier: Thermo Scientific Chemicals 228120010

Only null left
Add to Cart

CAS: 20830-75-5 | C41H64O14 | 780.94 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20830-75-5
Molecular Formula C41H64O14
Molecular Weight (g/mol) 780.94
MDL Number MFCD00003674
InChI Key LTMHDMANZUZIPE-PUGKRICDSA-N
Synonym digoxin, 12beta-hydroxydigitoxin, digoxine, lanoxin, lanoxicaps, digosin, digossina, digoxina, digoxinum, lanoxicaps tn
PubChem CID 2724385
ChEBI CHEBI:4551
IUPAC Name 3-[(3S,5R,8R,9S,10S,12R,13S,14S,17R)-3-[(2R,4S,5S,6R)-5-[(2S,4S,5S,6R)-5-[(2S,4S,5S,6R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tet
SMILES CC1C(C(CC(O1)OC2C(OC(CC2O)OC3C(OC(CC3O)OC4CCC5(C(C4)CCC6C5CC(C7(C6(CCC7C8=CC(=O)OC8)O)C)O)C)C)C)O)O

Specifications

Melting Point 248.0°C to 250.0°C
Assay Percent Range 94% min. (on dried substance) (HPLC)
Packaging Glass bottle
Quantity 1 g
Merck Index 15,3186
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in diluted alcohol,pyridine,mixture of,chloroform and alcohol,almost insoluble in ether,acetone,ethylacetate,,chloroform
Formula Weight 780.94
Percent Purity 95%
Chemical Name or Material Digoxin
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Fatal if swallowed.
Precautionary Statement GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
R
EINECSNumber 244-068-1

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.