Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Digoxin
GREENER_CHOICESDP

Catalog No. 458350 Shop All Novus Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-835-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 45-835-0 Supplier R&D Systems™ Supplier No. 4583/50
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Na+/K+ ATPase inhibitor

Inhibits membrane-bound α-subunits of the Na+/K+ ATPase pump in myocytes. Increases RGS2 protein levels in primary vascular smooth muscle cells through an Na+/K+ ATPase pump-dependent mechanism. Also exhibits anti-MERS-CoV activity in Vero cells in vitro (IC50 = 0.17 μM).

Chemical Identifiers

CAS 20830-75-5
Molecular Formula C41H64O14

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 780.94
Purity >98%
Molecular Weight (g/mol) 780.949
Chemical Name or Material Digoxin
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.