Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Digoxin Monoclonal Antibody (1H3)

Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA549705
Description
Antibody tested in indirect ELISA.
Digoxin is a beta blocker drug used to treat heart problems, such as heart failure, high blood pressure.
Specifications
| Digoxin | |
| Monoclonal | |
| 11 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Mouse | |
| Protein G | |
| RUO | |
| Chemical | |
| Antibody | |
| IgG |
| ELISA, Immunohistochemistry, Lateral Flow, SDS-Page | |
| 1H3 | |
| Unconjugated | |
| C41H64O14; DGX; Lanoxin | |
| Conjugate of DGX hapten and KLH. | |
| 200 μg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction