Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Digoxin Monoclonal Antibody (3301)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA127301
Description
MA1-27301 detects Digoxin.
Digoxin is a beta blocker drug used to treat heart problems, such as heart failure, high blood pressure.
Specifications
Digoxin | |
Monoclonal | |
5.78 mg/mL | |
0.015M potassium phosphate with 0.15M NaCl and 0.1% sodium azide; pH 7.2 | |
Mouse | |
Ion-exchange chromatography | |
RUO | |
Chemical | |
Antibody | |
IgG1 |
Radioimmune Assays (RIA) | |
3301 | |
Unconjugated | |
C41H64O14; DGX; Lanoxin | |
Digoxin, conjugated to BSA. | |
500 μg | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid | |
Chemical |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction