Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Digoxin Monoclonal Antibody (DG D8-20), Invitrogen™

Catalog No. 01671838
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-671-838 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-671-838 Supplier Thermo Scientific Supplier No. MA110810

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

MA1-10810 detects Digoxin. MA1-10810 has been successfully used in ELISA applications. The MA1-10810 immunogen is Digoxin, conjugated to KLH.

Digoxin is a beta blocker drug used to treat heart problems, such as heart failure, high blood pressure.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Digoxin
Applications ELISA
Classification Monoclonal
Clone DG D8-20
Concentration 1.46 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene Alias C41H64O14; DGX; Lanoxin
Host Species Mouse
Immunogen Digoxin, conjugated to KLH.
Purification Method purified
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Target Molecule Chemical
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.