Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Digoxin Monoclonal Antibody (DG D8-9), Invitrogen™

Catalog No. p-7245426
Change view
Click to view available options
Quantity:
1 mg
100 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA546140 100 μg
01-671-837 500 μg
PIMA546171 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. PIMA546140 Supplier Invitrogen™ Supplier No. MA546140
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

MA1-10809 detects Digoxin. MA1-10809 has been successfully used in ELISA applications. The MA1-10809 immunogen is Digoxin, conjugated to KLH.

Digoxin is a beta blocker drug used to treat heart problems, such as heart failure, high blood pressure.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Digoxin
Applications ELISA
Classification Monoclonal
Clone DG D8-9
Concentration 0.80 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene Alias C41H64O14; DGX; Lanoxin
Host Species Mouse
Immunogen Digoxin, conjugated to KLH.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Target Molecule Chemical
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.