Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Digoxin Monoclonal Antibody (DGD46), QED Bioscience™
SDP

Catalog No. 502277879
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-227-7879 1 mg
50-227-7881 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 50-227-7879 Supplier Leinco Technologies Inc Supplier No. 191031000UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Digoxin Monoclonal antibody specifically detects Digoxin in Chemical samples. It is validated for ELISA

Specifications

Antigen Digoxin
Applications ELISA
Classification Monoclonal
Clone DGD46
Concentration 0.5-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene Alias C41H64O14; DGX; Lanoxin
Host Species Mouse
Immunogen Digoxin-KLH.
Purification Method Protein A
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Target Molecule Chemical
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.