Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Digoxin Recombinant Rabbit Monoclonal Antibody (7H3V0)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA542751
This item is not returnable.
View return policy
Description
Immunogen sequence: Digoxin co njugated t o KLH.
Digoxin is a beta blocker drug used to treat heart problems, such as heart failure, high blood pressure.
Specifications
Digoxin | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
Chemical, Human, Mouse, Rat | |
Antibody | |
IgG |
Dot Blot, ELISA | |
7H3V0 | |
Unconjugated | |
C41H64O14; DGX; Lanoxin | |
Chemical compounds corresponding to Digoxin. | |
100 μL | |
Primary | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid | |
Chemical |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction