Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DIGRabbit anti-Plant, Clone: DIG44, Rockland™
SDP

Catalog No. 501991532
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-1532 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-199-1532 Supplier Rockland Immunochemicals Supplier No. 400001MG3
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. Expect reactivity against DIG-labeled probes and proteins.

Digoxin, also known as digitalis, is a purified cardiac glycoside extracted from the foxglove plant, Digitalis lanata. Its corresponding aglycone is digoxigenin, and its acetyl derivative is acetyldigoxin. Digoxin is widely used in the treatment of various heart conditions, namely atrial fibrillation, atrial flutter and sometimes heart failure that cannot be controlled by other medication. Digoxin preparations are commonly marketed under the trade names Lanoxin, Digitek, and Lanoxicaps. Digoxin is also used as an affinity system for nonradioactive labeling and detection of proteins and nucleic acids.

Specifications

Antigen DIG
Applications ELISA, Western Blot
Clone DIG44
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 25% glycerol and 0.01% sodium azide; pH 7.4
Gene Alias C23H34O5; DIG; Digitalis
Host Species Rabbit
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Plant
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.