Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(+)-Dihydrocarvone, 98%, mixture of isomers, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
5524-05-0
C10H16O
152.24
MFCD00001636
AZOCECCLWFDTAP-RKDXNWHRSA-N
+-dihydrocarvone,d-dihydrocarvone,1r,4r-dihydrocarvone,e-dihydrocarvone,unii-vta43h364z,2r-trans-2-methyl-5-1-methylvinyl cyclohexan-1-one,2r,5r-2-methyl-5-isopropenylcyclohexanone,2r,5r-5-isopropenyl-2-methylcyclohexanone,2r,5r-2-methyl-5-prop-1-en-2-yl cyclohexanone,cyclohexanone, 2-methyl-5-1-methylethenyl-, 2r,5r
22227
CHEBI:154
(2R,5R)-2-methyl-5-prop-1-en-2-ylcyclohexan-1-one
CC1CCC(CC1=O)C(=C)C
5524-05-0
C10H16O
152.24
MFCD00001636
AZOCECCLWFDTAP-RKDXNWHRSA-N
+-dihydrocarvone,d-dihydrocarvone,1r,4r-dihydrocarvone,e-dihydrocarvone,unii-vta43h364z,2r-trans-2-methyl-5-1-methylvinyl cyclohexan-1-one,2r,5r-2-methyl-5-isopropenylcyclohexanone,2r,5r-5-isopropenyl-2-methylcyclohexanone,2r,5r-2-methyl-5-prop-1-en-2-yl cyclohexanone,cyclohexanone, 2-methyl-5-1-methylethenyl-, 2r,5r
22227
CHEBI:154
(2R,5R)-2-methyl-5-prop-1-en-2-ylcyclohexan-1-one
CC1CCC(CC1=O)C(=C)C
Infrared Spectrum Authentic
Specific Gravity 0.9255
Refractive Index 1.4700 to 1.4740
Density 0.9255g/mL
Specific Rotation Condition + 14.00 (25.00°C c=neat)
Packaging Glass bottle
Boiling Point 87.0°C to 88.0°C (6.0mmHg)
Color Undesignated
Flash Point 81°C
Quantity 100g
Specific Rotation + 14.00
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Formula Weight 152.24
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material (+)-Dihydrocarvone, 98%
Show More Show Less

EINECSNumber : 226-872-4

TSCA : TSCA