Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dihydrofluorescein diacetate, 97%

Catalog No. AAJ6575903
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6575903 1 g
AAJ65759MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6575903 Supplier Thermo Scientific Chemicals Supplier No. J6575903
Add to Cart

Fluorescent dye for detecting intracellular oxidants | CAS: 35340-49-9 | C24H18O7 | 418.401 g/mol

Fluorescent dye for detecting intracellular oxidants

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35340-49-9
Molecular Formula C24H18O7
Molecular Weight (g/mol) 418.401
MDL Number MFCD00005056
InChI Key YKSJJXGQHSESKB-UHFFFAOYSA-N
Synonym dihydrofluorescein diacetate, 2-3,6-diacetoxy-9h-xanthen-9-yl benzoic acid, diacetyldihydrofluorescein, 2-3,6-diacetoxy-9h-xanth-9-yl benzoic acid, 2-3,6-bis acetyloxy-9h-xanthen-9-yl benzoic acid, dihydrofluorescein diacetate h2fda, 2-3,6-diacetyloxy-9h-xanthen-9-yl benzoic acid, 2-3,6-bis acetyloxy-9h-xanthen-9-yl benzoic acid #
PubChem CID 629056
IUPAC Name 2-(3,6-diacetyloxy-9H-xanthen-9-yl)benzoic acid
SMILES CC(=O)OC1=CC2=C(C=C1)C(C3=C(O2)C=C(C=C3)OC(=O)C)C4=CC=CC=C4C(=O)O

Specifications

Melting Point 213°C to 215°C
Color White to Yellow
Quantity 1 g
Beilstein 364167
Formula Weight 418.39
Percent Purity 97%
Physical Form Powder
Chemical Name or Material Dihydrofluorescein diacetate
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.