Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dihydrofolate Reductase/DHFR Mouse anti-Human, PerCP, Clone: 5B2, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259464PCP
This item is not returnable.
View return policy
Description
Dihydrofolate Reductase/DHFR Monoclonal antibody specifically detects Dihydrofolate Reductase/DHFR in Human samples. It is validated for Flow CytometrySpecifications
Dihydrofolate Reductase/DHFR | |
Monoclonal | |
PerCP | |
DHFRP1, dihydrofolate reductase, DYR, EC 1.5.1.3 | |
Recombinant human Dihydrofolate Reductase/DHFR (1-187aa) purified from E. coli | |
0.1 mL | |
Cancer | |
1719 | |
Store at 4°C in the dark. | |
IgG1 κ |
Flow Cytometry | |
5B2 | |
PBS | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction