Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dihydrofolate Reductase/DHFR Rabbit anti-Human, Mouse, Rat, Clone: 7N3X4, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | Dihydrofolate Reductase/DHFR |
---|---|
Clone | 7N3X4 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
Dihydrofolate Reductase/DHFR Monoclonal antibody specifically detects Dihydrofolate Reductase/DHFR in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
Dihydrofolate Reductase/DHFR | |
Western Blot | |
Unconjugated | |
Rabbit | |
Cancer | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
1719 | |
IgG | |
Affinity purified |
7N3X4 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
DHFRP1, dihydrofolate reductase, DYR, EC 1.5.1.3 | |
A synthetic peptide corresponding to a sequence within amino acids 88-187 of human Dihydrofolate Reductase/DHFR (P00374). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title