Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dihydrogen hexachloroplatinate(IV) hexahydrate, ACS, Premion™, 99.95% (metals basis), Pt 37.5% min

Catalog No. AA3625906
Click to view available options
Quantity:
1 g
5 g
25 g

Catalog No. AA3625906

Supplier: Thermo Scientific Chemicals 03625906

Only null left
Add to Cart

CAS: 18497-13-7 | Cl6H14O6Pt | 517.89 g/mol

Dihydrogen hexachloroplatinate(IV) hexahydrate is used for the determination of potassium. It is also used for the purification of platinum by treating it with ammonium chloride flowed by heating of ammonium hexachloroplatinate with hydrogen yields elemental platinum. It also finds application in catalysis, electroplating, photography, platinum mirrors, printing for etching of zinc, producing fine color in glass and porcelain, indelible ink and microscopy. It is a precursor for the preparation of other platinum catalysts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18497-13-7
Molecular Formula Cl6H14O6Pt
Molecular Weight (g/mol) 517.89
MDL Number MFCD00149910
InChI Key PIJUVEPNGATXOD-UHFFFAOYSA-J
IUPAC Name platinum(4+) dihydrogen hexahydrate hexachloride
SMILES [H+].[H+].O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Pt+4]

Specifications

Melting Point 60°C (decomposition)
Color Red to Brown
Physical Form Lump
Quantity 5 g
Assay Percent Range 99.95% (metals basis)
Solubility Information Soluble in water,ethanol,ether,ethyl acetate and acetone. Insoluble in nitric acid.
Formula Weight 517.91 (409.82 Anhydrous)
Grade ACS
Sensitivity Hygroscopic
Appearance Red-brown lump
Density 2.43
Chemical Name or Material Dihydrogen hexachloroplatinate(IV) hexahydrate
Show More Show Less
Hazard Category H301-H314-H317-H334-H335
Precautionary Statement P260-P264b-P270-P271-P272-P280-P285-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P501c
DOTInformation DOT Class: 8; Packing Group: III
EINECSNumber 241-010-7
RTECSNumber TP1500000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.